Learn More
NEOGEN Columbia Agar
SDP
Supplier:  NEOGEN 700003000
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Columbia Agar is used with or without blood for the isolation and cultivation of a wide variety of fastidious microorganisms. Conforms to Harmonized USP/EP/JP Requirements. Columbia Agar is not intended for use in the diagnosis of disease or other conditions in humans. A medium recommended by the Harmonized USP/EP/JP for isolation and identification of Clostridia from non-sterile products. Conforms to Harmonized USP/EP/JP performance specification. Originally described as a general purpose nutritious agar base by Ellner et al. at Columbia University that can be enriched by the addition of sterile blood. The peptone mixture and yeast extract provide a source of nitrogen, essential vitamins and amino acids. The starch provides a carbon source and sodium chloride maintains osmotic balance.For more information visit https://www.neogen.com/solutions/Microbiology/columbia-agar
Catalog No. 50-201-5189