Learn More
NEOGEN Columbia Blood Agar Base, 10 kg
Supplier: NEOGEN 700003055
Columbia Blood Agar Base is used with blood for the isolation and cultivation of a wide variety of fastidious microorganisms. Columbia Blood Agar Base is not intended for use in the diagnosis of disease or other conditions in humans. A modification of the original Columbia Agar base formulation, Columbia Blood Agar Base provides a medium that is suitable for use with both defibrinated horse or sheep blood. The peptone mixture provides a source of nitrogen, essential vitamins and amino acids. The starch provides a carbon source and sodium chloride maintains osmotic balance. Originally described as a general purpose nutritious agar base by Ellner et al. Columbia Agar is more frequently used when enriched by the addition of sterile blood. The medium is suitable for supporting the growth and determining hemolytic reactions of a variety of microorganisms. For more information visit https://www.neogen.com/solutions/Microbiology/columbia-blood-agar-base
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.