Learn More
Liofilchem™ ComASP™ Colistin Test Panel
ComASP™ Colistin is a 4-test panel containing the dried up antibiotic in 7 two-fold dilutions (0.25 to 16 µg/mL)
Supplier: Liofilchem™ 75001
Description
System is used to perform the broth microdilution (BMD) method for the antimicrobial susceptibility testing of colistin (polymyxin E) as recommended by international standards (i.e. CLSI, EUCAST, ISO) but in a simpler and less time-consuming way.
Kit Content:
- 4 Systems (panels) of ComASP Colistin panels individually packed in foil with silica gel desiccant)
- 16 Vials of Mueller Hinton II Broth (3.6 ml)
- Sealing Film
- Instructions Sheet and Test Results Form
Necessary items not included in the kit:
- McFarland 0.5 Barium Sulphate Standard (ref.80400) - Tips for multi.channel pipette (ref.96758)
- Solution reservoir for multichannel pipette (ref. 96761) - Multichannel pipette 30-300 µl (ref.96759)
- Physiological Solution (ref. 20095)
Specifications
Store ComASP Colistin at 2 to 8°C in the original packaging. Once an envelope is opened the panel should be used within 7 days. Keep away from direct sunlight and direct heat. | |
Used to perform the broth microdilution (BMD) method for the antimicrobial susceptibility testing of colistin (polymyxin E) as recommended by international standards (i.e. CLSI, EUCAST, ISO) but in a simpler and less time-consuming way. | |
4 Systems (panels) |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Non-hazardous according to current legislation but fall within the specific field of application where a safety data sheet must be supplied because they can cause phenomena of sensitisation in sensitive subjects if they come into contact with the skin.