Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ampco Safety Tools Combination Wrench, 2-1/2in.
SDP

Catalog No. 17777101 Shop All Ampco Metal Inc. Products
Click to view available options
:

Ampco Safety Tools Combination Wrench, 2-1/2in. is non-sparking, non-magnetic and corrosion resistant

Made in USA

Certifications

FM Approved

Warranty and Services

Lifetime Warranty with proper use. If a tool breaks or fails to perform under normal use aor service, it will be repaired or eplaced at no charge - exceptions include defects resulting from misuse, abuse or normal service wear.

Specifications

For Use With (Equipment)

Electronics, batteries, MRI, and precision magnetic equipment

Length (Metric)

577.85 mm

Material

Aluminum Bronze

Color

Bronze

For Use With (Application)

Non-Sparking, Non-Magnetic, Corrosion Resistant Applications; Flammable or combustible vapors, liquids, dusts or residues; Corrosive environments like laboratories, acid, salt water and chemical processing

No. per Pack

1 Ea.

Weight (Metric)

3630 g

Width (Metric)

117.47 mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.