Learn More
Bellco Glass, Inc. Bellco compression fittings are designed using our standard 45mm blue polypropylene open top caps and white silicone gaskets. A wide variety of fittings are available to allow ph and do monitoring via insertion of these probes into sidearms or vessels. Be
SDP
Supplier:  Bellco Glass, Inc. 196490115
Encompass_Preferred
Bellco compression fittings are designed using our standard 45mm blue polypropylene open top caps and white silicone gaskets. A wide variety of fittings are available to allow ph and do monitoring via insertion of these probes into sidearms or vessels. Be
Catalog No. 50-194-4934
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.