Learn More
Biotium Concanavalin A, CF640R Conjugate
Supplier: Biotium 29019
This product is fluorescently-labeled concanavalin A (Con A). Con A is a widely used lectin that selectively binds to a-mannopyranosyl and a-glucopyranosy residues, found in the cell wall of yeast and fungi, and the cell membrane of mammalian cells. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Far-red fluorescent CF640R dye has excitation/emission at 642/662 nm.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.