Learn More
Chemglass Life Sciences Condenser, Graham, 330mm, 24/40 Joint, 200mm Jacket Length
SDP
Supplier:  Chemglass Life Sciences CG121401
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Condenser, Graham, 330mm, 24/40 Joint, 200mm Jacket Length. Condenser has a top standard taper outer and lower inner drip tip joint. Hose connections have an O.D. of 10mm at the middle serration.
Catalog No. CG121401
missing translation for 'QuantityLeft'