Learn More
Ace Glass, Inc. Condenser, Graham, 200mm, 24/40
Supplier:  Ace Glass, Inc. 597712
Encompass_Preferred
200mm Graham condenser with 24/40 standard taper inner and outer joint at top and bottom. Hose connections are size D for 3/8 ID tubing.
Catalog No. 50-972-689
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.