Learn More
Ted Pella Inc Carbon Conductive Tape, 12mm W x 5m L
Supplier: Ted Pella Inc 160847
The conductive adhesive is a carbon–filled acrylic adhesive, free of solvents. It can be removed from the specimen mount with ethyl acetate, ethanol, isopropanol or alcohols. Temperature maximum is 60°C (140°F). Small impurities of Cu, Si, Sb, S, Na, P and very small impurities of Fe and Mg can be found. Refrigeration will increase shelf life but a warm-up time of 1 hour is then required before use.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.