Learn More
Cole-Parmer Oakton Conductivity and TDS Standard, 15,000 µS; 500 mL
Supplier: Cole-Parmer 0065350
All Oakton Conductivity and TDS Standards are traceable to NIST standard reference materials. The solutions help you calibrate conductivity instruments to ensure accuracy with standard values given at 25C. Use when readings are expected to be around 15 mS (15,000 S/cm).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.