Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Continental Copper Solution, Reagents
Continental Copper Solution, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$162.40 - $376.50
Specifications
Chemical Name or Material | Continental Copper Solution |
---|---|
Color | Blue |
CAS | 7732-18-5, 7758-99-8 |
Boiling Point | 100°C |
Specific Gravity | 1.1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
01-335-681
|
Reagents Holdings Llc
CC09000020F |
20 L |
Each for $376.50
|
|
|||||
01-335-680
|
Reagents Holdings Llc
CC0900004A |
4 L |
Each for $162.40
|
|
|||||
Specifications
Continental Copper Solution | |
7732-18-5, 7758-99-8 | |
1.1 | |
Non-sterile |
Blue | |
100°C | |
<1 | |
Liquid |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title