Learn More
Cole-Parmer Argos Technologies PolarSafe Aluminum Cooling Block, 15-Well for Cryogenic Vials
Supplier:  Cole-Parmer 6361502
Encompass_Preferred
Place blocks on ice, dry ice, liquid nitrogen, or in a water bath. These anodized aluminum blocks can keep samples at or near 1°C for up to 8 hours in properly packed ice, or for 3 to 7 hours in dry ice. Choose the blocks that meet your application needs — configurations are available for 1.5-/2.0-mL microcentrifuge tubes and cryogenic vials, 0.5-mL tubes, and 96-well or 364-well plates.
Catalog No. 50-196-0626
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.