Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Copper ICP Standard, 10,000 ppm Cu in 5% HNO3, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
50 mL
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
Specifications
Specifications
Packaging | Natural Poly Bottle |
Quantity | 50 mL |
CAS Min % | 88.0, 8.94, 0.88 |
CAS Max % | 92.0, 9.31, 0.92 |
Color | Blue |
Concentration or Composition Notes | 10,000ppm Cu |
Equivalence | 1 mL = 10 mg Cu |
Physical Form | Liquid |
Chemical Name or Material | Copper ICP Standard |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction