Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(II) 2,3,9,10,16,17,23,24-Octafluorophthalocyanine (purified by sublimation) 98.0+%, TCI America™
SDP

Catalog No. p-7125454 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
100 mg
1 g

Chemical Identifiers

CAS 148651-60-9
Molecular Formula C32H8CuF8N8
Molecular Weight (g/mol) 720.005
InChI Key SYOZEEBBXMOLOS-UHFFFAOYSA-N
Synonym 2,3,9,10,16,17,23,24-Octafluorophthalocyanine Copper(II), F8CuPc
PubChem CID 53384437
SMILES C1=C2C(=CC(=C1F)F)C3=NC4=NC(=NC5=NC(=NC6=C7C=C(C(=CC7=C([N-]6)N=C2[N-]3)F)F)C8=CC(=C(C=C85)F)F)C9=CC(=C(C=C94)F)F.[Cu+2]

Specifications

Color Blue-Purple
Quantity 100 mg
Formula Weight 720.01
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material Copper(II) 2,3,9,10,16,17,23,24-Octafluorophthalocyanine (purified by sublimation)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.