Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Copper(II) carbonate basic, Honeywell™
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
250g
1kg
Description
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Specifications
Specifications
Quantity | 250g |
Linear Formula | CuCO3 · Cu(OH)2 |
UN Number | NONH for all modes of transport |
Formula Weight | 221.12g/mol |
Percent Purity | ≥95% (RT) |
Chemical Name or Material | Copper(II) carbonate basic |
Safety and Handling
Hazard Statement | P261-P305 + P351 + P338 |
RTECSNumber | GL6910000 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction