Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Copper (II) Chloride, Dihydrate
SCI_ED

Catalog No. S25279B Shop All AquaPhoenix Scientific Products
Click to view available options
Quantity:
Packaging:
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS

10125-13-0

Molecular Formula

Cl2CuH4O2

Molecular Weight (g/mol)

170.48

MDL Number

MFCD00149674

InChI Key

MPTQRFCYZCXJFQ-UHFFFAOYSA-L

Synonym

coppertrace, copper ii chloride dihydrate, copper chloride dihydrate, caswell no. 235a, copper 2+ chloride dihydrate, cupric chloride usp, copper chloride cucl2 , dihydrate, epa pesticide chemical code 023701

PubChem CID

61482

ChEBI

CHEBI:86318

IUPAC Name

copper(2+) dihydrate dichloride

SMILES

O.O.[Cl-].[Cl-].[Cu++]

Specifications

Color

Blue-Green

Quantity

500 g

Linear Formula

CuCl2·2H2O

Formula Weight

170.48

Packaging

Lab Grade

Chemical Name or Material

Copper (II) Chloride, Dihydrate

ShelfLife

1 year

DOTInformation

DOT Class 8, III, : Corrosive

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.