Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Copper (II) Sulfate Pentahydrate, Reagent Grade
SCI_ED

Catalog No. S25286A Shop All AquaPhoenix Scientific Products
Click to view available options
Quantity:
Packaging:
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS

7758-99-8

Molecular Formula

CuH10O9S

Molecular Weight (g/mol)

249.68

MDL Number

MFCD00149681

InChI Key

JZCCFEFSEZPSOG-UHFFFAOYSA-L

Synonym

copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas

PubChem CID

24463

ChEBI

CHEBI:31440

IUPAC Name

copper(2+) pentahydrate sulfate

SMILES

O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

Specifications

Color

Blue

Quantity

500 g

Linear Formula

CuSO4·5H2O

Formula Weight

249.68

Packaging

Reagent Grade - Powder

Density

2.2840g/cm3

Chemical Name or Material

Copper (II) Sulfate, Pentahydrate, Reagent Grade

ShelfLife

2 years

DOTInformation

DOT Class 6.1, III, : Poison

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.