Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Copper Monoclonal Antibody (B4), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA549714

Description
Antibody tested in indirect ELISA
Copper is an essential trace mineral that plays a crucial role in many physiological processes in the human body. It is involved in the production of red blood cells, the maintenance of healthy bones and connective tissues, and the functioning of the nervous and immune systems. Copper is also a component of many enzymes, including those that are involved in energy production and antioxidant defense. The human body cannot produce copper on its own, so it must be obtained from dietary sources such as seafood, nuts, seeds, whole grains, and legumes. While copper deficiency is rare, excessive intake of copper can be toxic and lead to various health problems.Specifications
Copper | |
Monoclonal | |
1.9 mg/mL | |
PBS with 0.09% sodium azide, pH 7.4 | |
Mouse | |
Protein G | |
RUO | |
Mouse | |
Liquid |
ELISA, Immunohistochemistry, Lateral Flow, SDS-Page | |
B4 | |
Unconjugated | |
Cu | |
Conjugate of Cu hapten and KLH | |
200 μg | |
Primary | |
-20°, C, Avoid Freeze/Thaw Cycles | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction