Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Copper Standard, 2 ppm, Reagents
Click to view available options
:
Quantity: 4 L
Specifications
Specifications
Chemical Name or Material | Copper Standard, 2 ppm |
Packaging | Poly Bottle |
CAS | 7732-18-5, 7697-37-2 |
Quantity | 4 L |
Color | Colorless |
For Use With (Equipment) | Water Analyzer |
Grade | Reagent |
pH | <7 |
Boiling Point | 100°C |
Specific Gravity | 1 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction