Learn More
Corning™ Osteo Assay Surface Microplates
Capable of supporting the functional properties of osteogenic cells. Corning™ Osteo Assay Surface delivers lot-to-lot consistency, translating to reliable and reproducible results in bone cell assays. The surface also offers a consistent and defined alternative to preparing dentine or bone slices, reducing the variability in assay systems and resulting in more predictable assay readouts.
Supplier: Corning™ 3988
Description
- Individual alphanumerical codes for well identification, flat bottoms
- Treated for optimal cell attachment (except where noted)
- Corning™ CellBIND™ surface is a novel cell culture treatment that increases surface wettability for more even and consistent cell attachment.
- Ultra-Low Attachment surface plates feature a covalently bound hydrogel layer that minimizes cell attachment, protein absorption, and cellular activation.
- Corning Osteo Assay surface is an inorganic crystalline coating, creating a surface that mimics in vivo bone-like, for in vitro bone cell culture and assays.
- Sterile
- Nonpyrogenic
Specifications
With Lid | |
8 x 12 | |
Osteo Assay Surface | |
96-well Osteo Assay Surface Multiwell Microplate | |
360 μL | |
75 to 200 μL | |
Osteo Assay | |
Osteoblast and osteoclast differentiation and functional studies | |
1/Pk. | |
Clear |
0.32 cm2 | |
1/Pk; 4/Cs | |
Clear | |
Polystyrene | |
96 | |
Sterile | |
Round, Flat Bottom (F-well) | |
4/Cs. | |
4/Cs. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.