Learn More
AdipoGen Corynesidone A
SDP
Supplier:  AdipoGen AGCN20496M001
Encompass_Preferred
Chemical. CAS 1159583-61-5. Formula C15H12O5. MW 272.3. Isolated from Corynespora sp. Potent antioxidant. Reactive oxygen species ROS and reactive nitrogen species RNS scavenger. Anti-cancer and anti-inflammatory compound. Aromatase activity inhibitor. Moderate cytotoxic against selected cancer cell lines.
Catalog No. 50-256-2507
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.