Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coumarin 510 98.0+%, TCI America™
SDP

Catalog No. p-7125810 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
200 mg
1 g

Chemical Identifiers

CAS 87349-92-6
Molecular Formula C20H18N2O2
Molecular Weight (g/mol) 318.376
MDL Number MFCD00467042
InChI Key OEEAIIVRKIVRNX-UHFFFAOYSA-N
Synonym 2,3,6,7-Tetrahydro-10-(3-pyridyl)-1H,5H,11H-[1]benzopyrano[6,7,8-ij]quinolizin-11-one
PubChem CID 9905078
SMILES C1CC2=C3C(=C4C(=C2)C=C(C(=O)O4)C5=CN=CC=C5)CCCN3C1

Specifications

Melting Point 165°C
Color Yellow
Quantity 200 mg
Formula Weight 318.38
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Coumarin 510
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.