Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CPT 11, Tocris Bioscience™

Catalog No. p-4783884 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

DNA topoisomerase I inhibitor; antitumor

Inhibitor of DNA topoisomerase I that displays antitumor activity against a range of tumor types. Active metabolite available ( SN 38, Cat. No. 2684).

Chemical Identifiers

CAS 100286-90-6
Molecular Formula C33H39ClN4O6
Molecular Weight (g/mol) 623.15
MDL Number MFCD01862255
InChI Key GURKHSYORGJETM-WAQYZQTGSA-N
Synonym irinotecan hydrochloride, irinotecan hcl, topotecin, campto, cpt-11, cpt 11, camptothecin 11 hydrochloride, camptothecin 11, unii-06x131e4oe
PubChem CID 74990
ChEBI CHEBI:5971
IUPAC Name hydrogen (19S)-10,19-diethyl-19-hydroxy-14,18-dioxo-17-oxa-3,13-diazapentacyclo[11.8.0.0²,¹¹.0⁴,⁹.0¹⁵,²⁰]henicosa-1(21),2(11),3,5,7,9,15(20)-heptaen-7-yl [1,4'-bipiperidine]-1'-carboxylate chloride
SMILES [H+].[Cl-].CCC1=C2C=C(OC(=O)N3CCC(CC3)N3CCCCC3)C=CC2=NC2=C1CN1C2=CC2=C(COC(=O)[C@]2(O)CC)C1=O

Specifications

Quantity 10 mg
Formula Weight 623.14
Percent Purity >99%
Chemical Name or Material CPT 11
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.