Learn More
Electron Microscopy Sciences Fischione Carbon Film on Copper Tomography Grid
Supplier:  Electron Microscopy Sciences 7435704
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
This 1.5mm square, 300 mesh grid is made from copper and is for use with Fischione tomography specimen holders. The size allows for further tilt in TEMs with small pole piece holes. An identification mark and the square shape give easy reference when rotated past 90 deg.
Catalog No. 50-193-1249
missing translation for 'QuantityLeft'