Learn More
Electron Microscopy Sciences Square Grids, Material: Copper., 50/PK
Supplier:  Electron Microscopy Sciences CF150CUUL
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
A thin film of pure carbon coats the shiny side of a Gilder Finder grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Ultra-thin thickness is 3 to 4 nm. Material: Copper. Thickness: 3-4 nm .
Catalog No. 50-209-1153