Learn More
Electron Microscopy Sciences Carbon Support Film 5-6 nm thick on Square 200 mesh Nickel Grid
Supplier: Electron Microscopy Sciences CF200NI25
A thin film of pure carbon. All of our coated grids are optically checked followed by batch testing in the EM. Packed in grid storage box. NOTE: All of our film is laid on the shiny side of the grid. Standard thickness is 5-6 nm.
Grid: Nickel
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.