Learn More
Electron Microscopy Sciences Carbon Support Film 20-30 nm thick on Square 300 mesh Copper Grid
Supplier:  Electron Microscopy Sciences CF300CUET
Encompass_Preferred
A thin film of pure carbon. All of our coated grids are optically checked followed by batch testing in the EM. Packed in grid storage box. NOTE: All of our film is laid on the shiny side of the grid. Extra thick thickness is 20 to 30 nm.
Grid: Copper
Catalog No. 50-190-3083
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.