Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Creatinine, MP Biomedicals

Catalog No. p-7112283 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
10 g
25g
100g

  • Creatinine is a useful marker for normalizing levels of other molecules found in urine
  • Creatinine is a useful marker for normalizing levels of other molecules found in urine
  • Altered creatinine levels may be associated with conditions that result in decreased renal blood flow, such as diabetes and cardiovascular disease

Chemical Identifiers

CAS 60-27-5
Molecular Formula C4H7N3O
Molecular Weight (g/mol) 113.12
MDL Number MFCD00059730
InChI Key DDRJAANPRJIHGJ-UHFFFAOYSA-N
Synonym creatinine, 2-imino-1-methylimidazolidin-4-one, 1-methylglycocyamidine, 1-methylhydantoin-2-imide, creatinine,heated, 4h-imidazol-4-one, 2-amino-1,5-dihydro-1-methyl, 2-amino-1-methylimidazolin-4-one, creatinina, kreatinin, unii-ayi8ex34eu
PubChem CID 588
ChEBI CHEBI:16737
IUPAC Name 2-amino-3-methyl-4H-imidazol-5-one
SMILES CN1CC(=O)N=C1N

Specifications

Melting Point 295°C (literature)
Color White
Linear Formula C4H7N3O
Quantity 10 g
Solubility Information Soluble in 12 parts water; slightly soluble in ethanol; practically insoluble in acetone, ether, chloroform.
Formula Weight 113.1
Percent Purity 98.5 to 102.0%
Grade Anhydrous
Physical Form Powder
Chemical Name or Material CREATININE
Recommended Storage Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.