Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Creatinol Phosphate 98.0+%, TCI America™
SDP

Catalog No. p-7125642 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 6903-79-3
Molecular Formula C4H12N3O4P
Molecular Weight (g/mol) 197.13
MDL Number MFCD00868297
InChI Key FOIPWTMKYXWFGC-UHFFFAOYSA-N
Synonym 2-(1-Methylguanidino)ethyl Dihydrogen Phosphate
PubChem CID 23342
IUPAC Name [2-(N-methylcarbamimidamido)ethoxy]phosphonic acid
SMILES CN(CCOP(O)(O)=O)C(N)=N

Specifications

Melting Point 244°C
Color White
Boiling Point 244°C
Quantity 5 g
Formula Weight 197.13
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Creatinol Phosphate
RTECSNumber MF3600500
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.