Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cresyl Diphenyl Phosphate (so called) (mixture of analogue) 93.0+%, TCI America™
SDP

Catalog No. p-7134520 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 mL
500 mL

Chemical Identifiers

CAS 26444-49-5
Molecular Formula C19H17O4P
MDL Number MFCD00014920
Synonym Diphenyl Cresyl Phosphate, Phosphoric Acid Cresyl Diphenyl Ester

Specifications

Color Yellow
Boiling Point 390°C
Quantity 25 mL
UN Number 3082
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material Cresyl Diphenyl Phosphate (so called) (mixture of analogue)
EINECSNumber (2)-1860&(3)-2620
RTECSNumber TC5520000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.