Learn More
Kodiak™ Crusade CT WP 6 in. Black Boot
Offers a full-grain waterproof leather upper, waterproof membrane, an ASTM approved composite toe, slip and oil-resistant rubber outsole, and electrical hazard protection to help protect your feet, and confidently tackle tough days or terrain on the job-site.
Supplier: Kodiak™ K4NKBK 140 W
Description
- Full-grain waterproof leather upper with waterproof membrane construction
- KODIAK™ SOS Smell Out Science™ odor-control technology
- Abrasion-resistant leather toe guard offers added durability
- Full leather bellows tongue
- Removable, comfort trainer perforated EVA insole
- Kodiak™ Ultimate SR and oil-resistant rubber outsole with EVA midsole offers maximum comfort
Specifications
Black | |
14W | |
Yes | |
Boot |
Leather | |
Men's | |
Yes |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.