Learn More
United Scientific™ Cryo Coder For Skirted Base Vials
SDPSCI_ED
Easily identify different samples in your lab.
Supplier:  United Scientific™ P60104W
 View more versions of this product
Catalog No. S15764
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Description
Description
  • Designed to fit onto the top of our skirted base externally threaded cryogenic vial closures.
  • Provide quick visual identification of specific vials for ease in organizing inventory.
  • Available in a range of colors.
  • Compatible with United P60102 and P60103 skirted base cryo vials.
Specifications
Specifications
White
Cryo Coder
For Use with United Skirted Base Cryo Vials
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.