Learn More
United Scientific™ Cryo Coder For Star and Round Base Vials
SDPSCI_ED
Easily identify different samples in your lab.
Supplier:  United Scientific™ P60114R
 View more versions of this product
Catalog No. S15785
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Description
Description
  • Designed to fit onto the top of our star and round base cryogenic vial closures.
  • Provide quick visual identification of specific vials for ease in organizing inventory.
  • Available in a range of colors.
  • Compatible with all United Scientific cryo vials except P60102 and P60103 skirted base cryo vials.
Specifications
Specifications
Red
Cryo Coder
United Star and Round Base Cryo Vials
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.