Learn More
Electron Microscopy Sciences CryoCapsule High Pressure Freezing
Supplier: Electron Microscopy Sciences 3750
The CryoCapsule is used in High Pressure Freezing (HPF) and correlative light and electron microscopy (CLEM). Comparable to a small petri dish, it is composed of a landmarked sapphire disc and a gold spacer ring (50um thick) maintained together by a plastic ring. The specimens are encapsulated between the support sapphire disc (carbon landmarked) and a covering sapphire disc.
The CryoCapsule is loaded into a specific adaptor (HPM010, HPM100, HPF compact 02). Live cell imaging is done directly on the specimen in the CryoCapsule prior to HPF. Post-HPF, the specimen is processed for freeze substitution and room temperature sectioning.
CryoCapCell has developed a set of tools to manipulate the CryoCapsule in most scientific environments.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.