Learn More
Mitegen Crystal Loop 100Um B5(20) B
Supplier: Mitegen BM2100B5
MicroMount Assembly; box of 20 assemblies, 100 micron MicroMounts, 20 mounts on 18mm/SPINE length rods adhered into B5 bases.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.