Learn More
Hampton Research CRYSTALEX 96 WELL PLATE 10CSC
HR8-139/CrystalEX Second Generation 96 Well Plate (Corning 3552), 2 ul round drop well, 3 drop well, COC, untreated - 10 plate case. Please note items from this supplier are non-returnable.