Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CSM Media for Mav203 Yeast Cells


Click to view available options
:
2L
Description
The CSM Media for Mav203 Yeast Cells is a specifically designed media for making agar plates that enable the growth of Saccharomyces cerevisae MaV203 cells. This product comes with enough reagents to prepare 2 liters of solid media with the following components per liter:
Compatible with All MaV203 Applications
The CSM Media is optimized for the GeneArt™ High-Order Genetic Assembly System, including the recovery for transformations of vectors once they have been adapted to yeast using the GeneArt™ High-Order Vector Conversion Cassette. The media can also be used to grow MaV203 competent yeast cells, both library and sub-cloning scale (cat# 11281-011 and 11445-012).
Plate Preparation Protocol
For each liter, use one pouch of CSM -Trp Agar Yeast Media minus Glucose and 1 bottle of 20% Glucose. Dissolve the contents of the pouch in 900mL of distilled water and autoclave for 20 minutes on liquid cycle. Allow the solution to cool to 55°C and then add one bottle of 20% Glucose (100mL). Mix well and pour plates.
For Research Use Only. Not for use in diagnostic procedures.
- CSM (without tryptophan): 0.74g
- Yeast Nitrogen Base: 6.66g
- Ammonium Sulfate: 5g
- Glucose: 20g
- Agar: 20g
Compatible with All MaV203 Applications
The CSM Media is optimized for the GeneArt™ High-Order Genetic Assembly System, including the recovery for transformations of vectors once they have been adapted to yeast using the GeneArt™ High-Order Vector Conversion Cassette. The media can also be used to grow MaV203 competent yeast cells, both library and sub-cloning scale (cat# 11281-011 and 11445-012).
Plate Preparation Protocol
For each liter, use one pouch of CSM -Trp Agar Yeast Media minus Glucose and 1 bottle of 20% Glucose. Dissolve the contents of the pouch in 900mL of distilled water and autoclave for 20 minutes on liquid cycle. Allow the solution to cool to 55°C and then add one bottle of 20% Glucose (100mL). Mix well and pour plates.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Specifications
Content And Storage | CSM Media for MaV203 Yeast Cells has enough reagents to prepare 2 liters of agar media. The product contains two pouches of premixed powder and two 100 ml bottles of filter-sterilized, 20% glucose. Store all components at room temperature. All components are guaranteed stable for 6 months when properly stored. |
Form | Liquid, Powder |
Media Type | CSM Media |
Preparation Method | Autoclave |
Product Type | Microbiology Media |
Sterility | Non-sterile |
Target Organism Class | S. cerevisiae |
Final Product Type | Agar Plates |
For Use With (Application) | Optimized for use with the GENEART™ High-Order Genetic Assembly System, A13285 |
Product Line | GeneArt™ |
Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction