Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CU-76

Catalog No. 78202 Shop All R&D Systems Products
Click to view available options
:
2 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective inhibitor of cyclic GMP-AMP synthase (cGAS)

CU-76 is a selective cyclic GMP-AMP synthase (cGAS) inhibitor (IC50 = 0.24 μM). It specifically inhibits cGAS-STING pathway and has no effect on other nucleic acid sensing pathways such as the RIG-I-MAVS or Toll-like receptor pathways. CU-76 reduces DNA-induced IFN regulatory factor 3 (IRF3) dimerization in a dose-dependent manner, and it decreases production of IFN-β in THP-1 cells stimulated with interferon-stimulatory DNA.

Specifications

Chemical Name or Material Methyl 4-amino-6-[(3,5-difluoro-4-iodophenyl)amino]-1,3,5-triazine-2-carboxylate
CAS 2400954-58-5
Quantity 2 mg
Target STING/TMEM173
Molecular Formula C11H8F2IN5O2
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.