Learn More
Bioworld Cu-IMAC 5 x 2mL Columns, 5 Column
Supplier: Bioworld 201700091
Cu-immobilized metal affinity chromatography (IMAC) columns consist of Cu-Chelated Separoporematrix conveniently prepacked spin columns for gravity flow chromatography.Cu-IMAC Columns separate proteins via their differences in affinity toCu2+ metal ions. Ideal for purification of recombinant proteins containing (His)6 fusion tag.Technical SpecificationsImminodiacetate immobilized matrix charged withCu metal ions.Chelating group coupling method: epoxy activationMatrix: cross-linked Separopore 4BParticle size (range): 50-170µm Metal ion capacity: > 20µmol Cu2+/ml gelSpacer arm: 14 atompH stability: 3-12Binding Capacity: Approx. 40 mg His-tagged protein/ml mediumSupplied as suspension in 20% ethanol.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.