Learn More
Sklar Instruments Cube Pessary, Sklar, 37mm Cube Pessary With Drain, #3, Non-Sterile, Silicone, Latex Free. EA/1.
SDP
Supplier:  Sklar Instruments 905303
Encompass_Preferred
Cube Pessary, Sklar, 37mm Cube Pessary With Drain, #3, Non-Sterile, Silicone, Latex Free. Pessaries are OB/GYN products used for the non-surgical management of certain gynecologic conditions. There are a several types of pessaries, each designed and used for a specific treatment. This product is a #3, 37mm cube pessary with a dra in. It is used to treat moderate to severe cases of uterine prolapse as well as mild cystocele and rectocele. Insertion and removal of a cube pessary relies on suction to the vaginal walls and tends to be more difficult that other pessaries. This may cause patients discomfort. A cube pessary with a drain, such as this one, may be left inserted for up to a week before removal and cleaning is required. Cube pessaries without drains require daily removal and cleaning.
Catalog No. 50-143-6803
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.