Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brew Plate™ Custom - Whole plates of WLD
SCI_ED

Catalog No. S72041
Click to view available options
:

Brew Plate™ Custom - Whole plates of WLD is part of a collection of prefilled, easy-to-use beer testing kits and bulk plates that detect microbes that spoil beer during the brewing process.

  • The Brew Plate Bulk Sleeves offer sleeves of 25 plates that allow the brewer to detect a wide scope of microbes known to cause contamination during the brewing process.
  • Each plate provides sections with various media to detect common wild yeast, aerobic (lives with oxygen) and anaerobic bacteria (lives without oxygen).
  • If contamination is detected, samples or plates can be sent to the Brew Plate Laboratory for microbial identification. Each bulk sleeve purchase comes with one free sequence identification.

Specifications

Product Type

BrewPlate

For Use With (Application)

Cultivating yeast and bacteria found in brewing.

Content And Storage

Refrigeration

Manufacturing Origin

US

Quantity

25 Plates

Packaging Type

Bulk Sleeve

Media Type

Wallerstein Laboratories Differential Meda

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.