Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CY 208-243, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
Centrally active dopamine D1 receptor agonist, selective over D2 receptor sites. Stimulates adenylate cyclase in rat striatal homogenates with an EC50 of 125 nM. Unlike SKF 38393 (Cat. No. 0922), it exerts antiParkinsonian activity in animal models.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 274.36 |
Percent Purity | >98% |
Chemical Name or Material | CY 208-243 |
Safety and Handling
Recommended Storage | Store at Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction