Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyanogen iodide, 98%, Thermo Scientific Chemicals

Catalog No. p-2638680
Click to view available options
Quantity:
5 g
25 g

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 506-78-5
Molecular Formula CIN
InChI Key WPBXOELOQKLBDF-UHFFFAOYSA-N
Synonym cyanogen iodide, iodine cyanide, jodcyan, iodocyanide, iodine monocyanide, cyanogen monoiodide, iodine cyanide i cn, unii-81esv254uf, cyanic iodide, cyano iodide
PubChem CID 10478
IUPAC Name carbononitridic iodide
SMILES C(#N)I

Specifications

Melting Point 142.0°C to 149.0°C
Color Beige to Brown or Pink
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Iodometry)
Packaging Glass bottle
Linear Formula ICN
Quantity 5 g
Merck Index 15,2685
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in diethyl ether,ethanol,ether,alcohols,and volatile oils
Molecular Weight (g/mol) 152.92
Formula Weight 152.92
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Cyanogen iodide
Show More Show Less
Hazard Category GHS Signal Word: Danger
EINECSNumber 208-053-3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.