Learn More
American Radiolabeled Chemicals Inc CYCLIC AMP 3H 1LC
SDP
Supplier:  American Radiolabeled Chemicals Inc ART 17901MC
Encompass_Preferred
Adenosine 3',5'-cyclic monophosphate-5',8-3H ammonium salt 1mCi in ethanol:water (1:1), 1 mCi/ml, S.A. 60-90 Ci/mmol, M.W. 346.30, Shipped in Dry Ice, Exclusive
Catalog No. 50-901-218
missing translation for 'QuantityLeft'
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.