Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

γ- Cyclodextrin, MP Biomedicals™

Catalog No. p-4679717 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 17465-86-0
Molecular Formula C48H80O40
Molecular Weight (g/mol) 1297.13
MDL Number MFCD00009595,MFCD00149574
InChI Key GDSRMADSINPKSL-UHFFFAOYNA-N
Synonym cyclooctaamylose, schardinger gamma-dextrin
PubChem CID 101136809
IUPAC Name 5,10,15,20,25,30,35,40-octakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34,37,39-hexadecaoxanonacyclo[36.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹.2³³,³⁶]hexapentacontane-41,42,43,44,45,46,47,48,49,50,51,52,53,54,55,56-hexadecol
SMILES OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC9C(CO)OC(OC1C(O)C2O)C(O)C9O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O

Specifications

Quantity 1 g
Formula Weight 1297.1
Physical Form Solid
Chemical Name or Material γ- Cyclodextrin
Recommended Storage Store at room temperature.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.