Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopentylmalonic Acid 98.0+%, TCI America™
SDP

Catalog No. p-7124699 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 5660-81-1
Molecular Formula C8H12O4
Molecular Weight (g/mol) 172.18
MDL Number MFCD00059515
InChI Key DRHLPIHNSBNMFF-UHFFFAOYSA-N
PubChem CID 231700
IUPAC Name 2-cyclopentylpropanedioic acid
SMILES C1CCC(C1)C(C(=O)O)C(=O)O

Specifications

Melting Point 162°C
Color White
Quantity 5 g
Formula Weight 172.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Cyclopentylmalonic Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.