Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Cynomolgus Monkey CD38 (NP_001274206) VersaClone cDNA
Shop All R&D Systems Products
Click to view available options
:
10 μg
Description
The Cynomolgus Monkey CD38 (NP_001274206) VersaClone cDNA clone contains the complete ORF for CD38, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The CD38 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Specifications
Specifications
Gene | CD38 |
Accession Number | NP_001274206 |
Concentration | 10 μg at 0.2 μg/μL |
Formulation | cDNA is provided in 10 mM Tris-Cl |
Insert Type | 919bp |
pH | 8.5 |
Gene ID (Entrez) | 952 |
Gene Alias | ADP-ribosyl Cyclase, ADP-ribosyl cyclase 1, ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase, cADPr hydrolase 1, CD38, CD38 antigen, CD38 antigen (p45), CD38 molecule, Cyclic ADP-ribose Hydrolase, Cyclic ADP-ribose hydrolase 1, EC 3.2.2.5, NAD(+) nucleosidase, T10 |
Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
Species | Cynomolgus Monkey |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction