Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYP2A6 BACULOSOMES™ Plus Reagent, rHuman
SDP

Catalog No. PV6137
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
Click to view available options
:

CYP2A6 BACULOSOMES™ Plus Reagent, rHuman

Cytochrome P450 BACULOSOMES™ Plus Reagents are microsomes prepared from insect cells infected with recombinant baculovirus containing a human CYP450 isozyme, as well as human cytochrome P450 reductase. For this particular isozyme (CYP2A6), human cytochrome b5 is also included.

  • Single human P450 isozyme for detailed drug metabolism studies
  • High specific activity increases dynamic range and sensitivity
  • No background metabolism for clear results

    Single Overexpressed Human CYP450 Isozyme
    CYP450 BACULOSOMES™ Plus Reagents offer a distinct advantage over Human Liver Microsomes (HLMs) in that only one CYP450 isozyme is expressed, thereby preventing metabolism by other CYP450s or other classes of drug metabolizing enzymes.

    High Activity Results in Increased Sensitivity
    Compound rankings based on metabolism and inhibition profiles observed with CYP450 BACULOSOMES™ Plus Reagents are very similar to those seen with HLMs for most compounds tested. However, CYP450 BACULOSOMES™ Plus Reagents activity and metabolic rates with most substrates is significantly higher than those seen with HLMs. This results in a broad dynamic range and high sensitivity in assays utilizing CYP450 BACULOSOMES™ Plus Reagents. The high level of activity and reproducibility of the enzyme component observed in the reaction with most probe substrates makes CYP450 BACULOSOMES™ Plus Reagents well suited to high-throughput screening formats.

    High Signal to Noise Ratio
    No background metabolism by endogenous insect CYP450s has been detected with any of the Vivid™ probe substrates tested so far.

    Applications: isozyme identification, DMPK analysis, isozyme-specific metabolism, and inhibition screening

    For Research Use Only. Not for any animal or human therapeutic or diagnostic use.
  • Specifications

    Product Line

    BACULOSOMES™

    For Use With (Equipment)

    Microplate Reader

    Target

    Cytochrome P450

    Content And Storage

    1 tube, Store at -80 °C

    Product Type

    Bacterial Microsome

    Quantity

    0.5 nmol

    Format

    Liquid suspension

    Expression System

    Baculovirus

    Protein Subtype

    Cytochrome P450 Enzymes

    Shipping Condition

    Dry Ice

    Sub Cellular Localization

    Microsomes

    Research Category

    Toxicology & Drug Metabolism, Drug Development

    Species

    Human

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.