Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cystamine dihydrochloride, min. 97.0%, MP Biomedicals™

Catalog No. ICN10049201 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:

Chemical Identifiers

CAS

56-17-7

Molecular Formula

C4H14Cl2N2S2

Molecular Weight (g/mol)

225.19

InChI Key

YUFRRMZSSPQMOS-UHFFFAOYSA-N

Synonym

cystamine dihydrochloride, cystamin dihydrochloride, ethanamine, 2,2'-dithiobis-, dihydrochloride, usaf cb-34, 2,2'-disulfanediyldiethanamine dihydrochloride, 2-aminoethyl disulfide dihydrochloride, bis 2-aminoethyl disulfide dihydrochloride, cystamine 2hcl, 2,2'-dithiodi ethylammonium dichloride, decarboxycystine dihydrochloride

PubChem CID

5941

IUPAC Name

2-(2-aminoethyldisulfanyl)ethanamine;dihydrochloride

SMILES

C(CSSCCN)N.Cl.Cl

Specifications

Color

White

Assay Percent Range

min. 97.0%

Quantity

1 g

Formula Weight

225.2

Physical Form

Powder

Chemical Name or Material

Cystamine dihydrochloride

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.