Learn More
Medchemexpress LLC HY-N3686 1g Medchemexpress, D-Arabitol CAS:488-82-4 Purity:>98%
Supplier: Medchemexpress LLC HYN36861G
Medchemexpress, HY-N3686 1g D-Arabitol CAS:488-82-4 D-Arabitol is a polyol and its accumulation may cause a neurotoxic effect in human. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.